ENA245486788 Enamine

8-methyl-decahydroquinoline

  • MDL number MFCD09941178
  • Cas Number : 52601-69-1
  • Molecular Weight : 153.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54599
Availability details for
Quantity
  • Availability
  • Add to Cart