ENA245486789 Enamine

(1S)-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-ol

  • MDL number MFCD09863687
  • Cas Number : 132486-27-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54600
Availability details for
Quantity
  • Availability
  • Add to Cart