ENA245486790 Enamine

[1-(4-fluorophenyl)pyrrolidin-3-yl]methanamine

  • MDL number MFCD09945302
  • Cas Number : 933712-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54601
Availability details for
Quantity
  • Availability
  • Add to Cart