ENA245486799 Enamine

3-[4-(propan-2-yloxy)phenyl]propan-1-ol

  • MDL number MFCD08693169
  • Cas Number : 365572-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53797
Availability details for
Quantity
  • Availability
  • Add to Cart