ENA245486805 Enamine

6-(4-chlorophenoxy)pyridin-3-amine

  • MDL number MFCD00052646
  • Cas Number : 75926-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54633
Availability details for
Quantity
  • Availability
  • Add to Cart