ENA245486808 Enamine

1-[1-(3-bromophenyl)pyrrolidin-3-yl]methanamine

  • MDL number MFCD09939797
  • Cas Number : 1017043-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54636
Availability details for
Quantity
  • Availability
  • Add to Cart