ENA245486810 Enamine

methyl 2-{bicyclo[2.2.1]heptan-2-yl}acetate

  • MDL number MFCD13593679
  • Cas Number : 54441-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54638
Availability details for
Quantity
  • Availability
  • Add to Cart