ENA245486811 Enamine

(2-chloro-4-methoxyphenyl)(phenyl)methanone

  • MDL number MFCD12434896
  • Cas Number : 67601-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-54639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54639
Availability details for
Quantity
  • Availability
  • Add to Cart