ENA245486818 Enamine

1-(4-chlorophenyl)-2-phenylethan-1-amine hydrochloride

  • MDL number MFCD13195943
  • Cas Number : 92252-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-53801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53801
Availability details for
Quantity
  • Availability
  • Add to Cart