ENA245486819 Enamine

2-(2-methoxyethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09817191
  • Cas Number : 247582-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53802
Availability details for
Quantity
  • Availability
  • Add to Cart