ENA245486823 Enamine

2-(3,5-dimethylpiperidin-1-yl)-2-methylpropan-1-amine

  • MDL number MFCD09931617
  • Cas Number : 1016686-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53804
Availability details for
Quantity
  • Availability
  • Add to Cart