ENA245486827 Enamine

1-{5-[(diethylamino)methyl]-1,2,4-oxadiazol-3-yl}cyclopentan-1-amine dihydrochloride

  • MDL number MFCD13196107
  • Cas Number : 1208416-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54661
Availability details for
Quantity
  • Availability
  • Add to Cart