ENA245486832 Enamine

2-(methylamino)propanenitrile

  • MDL number MFCD00045617
  • Cas Number : 16752-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54666

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-54666
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart