ENA245486833 Enamine

8-chloro-5-methyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11651650
  • Cas Number : 22406-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53808
Availability details for
Quantity
  • Availability
  • Add to Cart