ENA245486838 Enamine

2-fluoro-6-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD07368711
  • Cas Number : 887266-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54672
Availability details for
Quantity
  • Availability
  • Add to Cart