ENA245486840 Enamine

1-cyanocyclohexane-1-carboxylic acid

  • MDL number MFCD02317148
  • Cas Number : 227203-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54675
Availability details for
Quantity
  • Availability
  • Add to Cart