ENA245486842 Enamine

(2,5-dimethoxyphenyl)(phenyl)methanone

  • MDL number MFCD00769055
  • Cas Number : 4038-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54677
Availability details for
Quantity
  • Availability
  • Add to Cart