ENA245486846 Enamine

3-[2-(2,4-dimethoxyphenyl)-1H-indol-3-yl]propanoic acid

  • MDL number MFCD09040962
  • Cas Number : 926259-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54683
Availability details for
Quantity
  • Availability
  • Add to Cart