ENA245486847 Enamine

4-chloro-2-(pyrrolidin-1-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD13188364
  • Cas Number : 1211296-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54691
Availability details for
Quantity
  • Availability
  • Add to Cart