ENA245486850 Enamine

6,12-dibromo-1,4-dioxaspiro[4.7]dodecane

  • MDL number MFCD00795430
  • Cas Number : 3514-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54694
Availability details for
Quantity
  • Availability
  • Add to Cart