ENA245486853 Enamine

2-[4-(trifluoromethoxy)phenyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD12068247
  • Cas Number : 1157562-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54699
Availability details for
Quantity
  • Availability
  • Add to Cart