ENA245486856 Enamine

methyl 3-amino-2-[(4-fluorophenyl)methyl]propanoate hydrochloride

  • MDL number MFCD13196115
  • Cas Number : 1209305-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54702
Availability details for
Quantity
  • Availability
  • Add to Cart