ENA245486857 Enamine

1-(prop-2-en-1-yl)-1H-indole-3-carboxylic acid

  • MDL number MFCD11155405
  • Cas Number : 1021244-66-5
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54703
Availability details for
Quantity
  • Availability
  • Add to Cart