ENA245486860 Enamine

5-methoxy-2-nitro-4-[(oxolan-2-yl)methoxy]benzoic acid

  • MDL number MFCD13196117
  • Cas Number : 1096877-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54707
Availability details for
Quantity
  • Availability
  • Add to Cart