ENA245486875 Enamine

N-(2-aminoethyl)-N-methylcyclopropanamine

  • MDL number MFCD11623815
  • Cas Number : 126105-24-6
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54723
Availability details for
Quantity
  • Availability
  • Add to Cart