ENA245486881 Enamine

1-(4-methoxyphenyl)-1-(4-methylphenyl)methanamine hydrochloride

  • MDL number MFCD08276233
  • Cas Number : 1067902-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54738
Availability details for
Quantity
  • Availability
  • Add to Cart