ENA245486883 Enamine

1-(methylamino)-3-[2-(trifluoromethyl)phenoxy]propan-2-ol

  • MDL number MFCD11135144
  • Cas Number : 1019477-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53813
Availability details for
Quantity
  • Availability
  • Add to Cart