ENA245486885 Enamine

4-methoxy-3-methylbenzene-1-sulfonamide

  • MDL number MFCD03197280
  • Cas Number : 84910-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54747
Availability details for
Quantity
  • Availability
  • Add to Cart