ENA245486888 Enamine

1-amino-3-[4-(trifluoromethyl)phenoxy]propan-2-ol

  • MDL number MFCD10691815
  • Cas Number : 858619-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54750
Availability details for
Quantity
  • Availability
  • Add to Cart