ENA245486889 Enamine

2-[2-(5-chlorothiophen-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD11102968
  • Cas Number : 1152605-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54752
Availability details for
Quantity
  • Availability
  • Add to Cart