ENA245486892 Enamine

3-[4-(trifluoromethyl)phenyl]butanoic acid

  • MDL number MFCD11172905
  • Cas Number : 1042815-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54755

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54755
Availability details for
Quantity
  • Availability
  • Add to Cart