ENA245486893 Enamine

5-oxo-3-phenyl-5H-pyrimido[2,1-b][1,3]thiazole-6-carboxylic acid

  • MDL number MFCD03011515
  • Cas Number : 446829-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54756
Availability details for
Quantity
  • Availability
  • Add to Cart