ENA245486896 Enamine

{3-[(1,3-dioxaindan-5-yl)methoxy]-2-hydroxypropyl}(methyl)amine hydrochloride

  • MDL number MFCD13196126
  • Cas Number : 1210154-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54759
Availability details for
Quantity
  • Availability
  • Add to Cart