ENA245486903 Enamine

ethyl 4-(2,4-dimethylphenyl)-2,4-dioxobutanoate

  • MDL number MFCD03411531
  • Cas Number : 1019340-49-8
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54768
Availability details for
Quantity
  • Availability
  • Add to Cart