ENA245486904 Enamine

2-(cyclopropylmethoxy)benzaldehyde

  • MDL number MFCD11156430
  • Cas Number : 54280-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54769
Availability details for
Quantity
  • Availability
  • Add to Cart