ENA245486905 Enamine

3-[(4-bromophenyl)sulfanyl]propanoic acid

  • MDL number MFCD05664871
  • Cas Number : 13735-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53711

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-53711
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart