ENA245486909 Enamine

2,3-dimethylbutan-2-amine hydrochloride

  • MDL number MFCD01694856
  • Cas Number : 29772-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292-293
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54776
Availability details for
Quantity
  • Availability
  • Add to Cart