ENA245486910 Enamine

2-chloro-6-nitrobenzene-1-sulfonamide

  • MDL number MFCD13196131
  • Cas Number : 89581-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54777
Availability details for
Quantity
  • Availability
  • Add to Cart