ENA245486911 Enamine

2-chloro-6-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD13196132
  • Cas Number : 17970-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54778
Availability details for
Quantity
  • Availability
  • Add to Cart