ENA245486913 Enamine

methyl 2-amino-5-phenoxybenzoate

  • MDL number MFCD13196134
  • Cas Number : 1100393-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54781

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54781
Availability details for
Quantity
  • Availability
  • Add to Cart