ENA245486919 Enamine

methyl 4-aminopiperidine-1-carboxylate

  • MDL number MFCD11128922
  • Cas Number : 1019351-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54790
Availability details for
Quantity
  • Availability
  • Add to Cart