ENA245486920 Enamine

1-(3-bromopropanesulfonyl)-4-fluorobenzene

  • MDL number MFCD11164525
  • Cas Number : 1082893-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54791
Availability details for
Quantity
  • Availability
  • Add to Cart