ENA245486921 Enamine

5-[(diethylamino)methyl]furan-2-carbohydrazide

  • MDL number MFCD06347512
  • Cas Number : 91141-17-2
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54792
Availability details for
Quantity
  • Availability
  • Add to Cart