ENA245486928 Enamine

2-chloro-6-(3,5-dimethyl-1H-pyrazol-1-yl)pyrazine

  • MDL number MFCD11621999
  • Cas Number : 1133082-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54813
Availability details for
Quantity
  • Availability
  • Add to Cart