ENA245486929 Enamine

N-(3-carbamothioylphenyl)-2-phenylbutanamide

  • MDL number MFCD09929967
  • Cas Number : 1017090-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54814
Availability details for
Quantity
  • Availability
  • Add to Cart