ENA245486930 Enamine

2-amino-N-cyclopentyl-3-methylbutanamide

  • MDL number MFCD09943996
  • Cas Number : 1103225-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54816
Availability details for
Quantity
  • Availability
  • Add to Cart