ENA245486936 Enamine

hexyl[2-(morpholin-4-yl)ethyl]amine

  • MDL number MFCD11180297
  • Cas Number : 100392-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54822
Availability details for
Quantity
  • Availability
  • Add to Cart