ENA245486946 Enamine

2-(2,6-dioxopiperidin-1-yl)acetic acid

  • MDL number MFCD10007769
  • Cas Number : 147104-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54837
Availability details for
Quantity
  • Availability
  • Add to Cart