ENA245486947 Enamine

N-(1-ethyl-1H-pyrazol-4-yl)piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD13196140
  • Cas Number : 1210224-02-4
  • Molecular Weight : 295.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54839
Availability details for
Quantity
  • Availability
  • Add to Cart