ENA245486950 Enamine

(2,2-difluoroethyl)(methyl)amine hydrochloride

  • MDL number MFCD11043069
  • Cas Number : 139364-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54843

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-54843
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart